Gene Transcripts

Gene Symbol
Ezr
Entrez Gene ID
54319
Gene Name
ezrin
Gene Aliases
Vil2
Chromosome Location
Chr. 1: 47287872 - 47331412 on Build Rnor_6.0
UniGene ID
Rn.773
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019357.1 NP_062230.1 2, 3 586
GenBank mRNA AF450298.1 - 2, 3 185
AY428869.1 - 2, 3 171
BC081958.1 - 2, 3 586 7124292

View Full Product Details