Gene Transcripts

Gene Symbol
Tusc1
Entrez Gene ID
69136
Gene Name
tumor suppressor candidate 1
Gene Aliases
2200001D17Rik, TSG-9
Chromosome Location
Chr. 4: 93334148 - 93335511 on Build GRCm38
UniGene ID
Mm.171558
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026954.1 NP_081230.1 1 793
GenBank mRNA AK008612.1 - 1 504
AY546089.1 - 1 793

View Full Product Details