Gene Transcripts

Gene Symbol
Lyrm7
Entrez Gene ID
75530
Gene Name
LYR motif containing 7
Gene Aliases
1700024C24Rik, 9330147L21Rik
Chromosome Location
Chr. 11: 54826866 - 54860907 on Build GRCm38
UniGene ID
Mm.83578
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029327.3 NP_083603.2 5 473
GenBank mRNA AK006294.1 - 5 800
BC139403.1 - 5 604 8862425
BC139406.1 - 5 604 8862428

View Full Product Details