Gene Transcripts

Gene Symbol
Tmem35
Entrez Gene ID
308134
Gene Name
transmembrane protein 35
Gene Aliases
RSEP4
Chromosome Location
Chr. X: 105134498 - 105145346 on Build Rnor_6.0
UniGene ID
Rn.45854
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001799.1 NP_001001799.1 2 1155
GenBank mRNA AY540309.1 - 2 1155

View Full Product Details