Gene Transcripts

Gene Symbol
NAV3
Entrez Gene ID
89795
Gene Name
neuron navigator 3
Gene Aliases
POMFIL1, STEERIN3, unc53H3
Chromosome Location
Chr. 12: 77571862 - 78213012 on Build GRCh38
UniGene ID
Hs.655301
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024383.1 NP_001019554.1 8 1599
NM_014903.5 NP_055718.4 8 1599
XM_005269215.3 XP_005269272.1 8 5600
XM_011538940.2 XP_011537242.1 8 5601
XM_011538941.2 XP_011537243.1 8 5600
XM_011538944.2 XP_011537246.1 8 5598
XM_017020164.1 XP_016875653.1 8 5602
XM_017020165.1 XP_016875654.1 8 5602
XM_017020166.1 XP_016875655.1 8 1588
XM_017020167.1 XP_016875656.1 7 1519
XM_017020168.1 XP_016875657.1 8 5601
XM_017020169.1 XP_016875658.1 8 5600
XM_017020170.1 XP_016875659.1 8 5597
GenBank mRNA AF397731.1 - 7 1183
AJ488201.1 - 8 1471
BC017667.2 - 8 1976

View Full Product Details