Gene Transcripts

Gene Symbol
DPPA3
Entrez Gene ID
359787
Gene Name
developmental pluripotency associated 3
Gene Aliases
STELLA
Chromosome Location
Chr. 12: 7711454 - 7717559 on Build GRCh38
UniGene ID
Hs.131358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199286.3 NP_954980.1 3 476
GenBank mRNA AY317075.1 - 3 476

Gene Symbol
DPPA3P2
Entrez Gene ID
400206
Gene Name
developmental pluripotency associated 3 pseudogene 2
Gene Aliases
STELLAR
Chromosome Location
Chr. 14: 36371318 - 36372369 on Build GRCh38
UniGene ID
Hs.528118
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY230136.1 - 1 415
BC062480.1 - 1 608 30398001
BC125145.1 - 1 427 40120129

View Full Product Details