Gene Transcripts

Gene Symbol
Kif5a
Entrez Gene ID
314906
Gene Name
kinesin family member 5A
Gene Aliases
Chromosome Location
Chr. 7: 70515832 - 70552897 on Build Rnor_6.0
UniGene ID
Rn.63144
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212523.1 NP_997688.1 9 958
GenBank mRNA AY535015.1 - 9 958
FQ212220.1 - 998

View Full Product Details