Gene Transcripts

Gene Symbol
BOLA3
Entrez Gene ID
388962
Gene Name
bolA family member 3
Gene Aliases
MMDS2
Chromosome Location
Chr. 2: 74135401 - 74147912 on Build GRCh38
UniGene ID
Hs.61472
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035505.1 NP_001030582.1 2 182
NM_212552.2 NP_997717.2 2 182
GenBank mRNA BC028282.1 - 2 142
BC042036.1 - 2 178
BU600037.1 - 187
DQ225187.1 - 2 144

View Full Product Details