Gene Transcripts

Gene Symbol
Nlrp9b
Entrez Gene ID
243874
Gene Name
NLR family, pyrin domain containing 9B
Gene Aliases
Nalp-delta, Nalp9b, Nlrp9
Chromosome Location
Chr. 7: 19991393 - 20062942 on Build GRCm38
UniGene ID
Mm.277114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194058.2 NP_918947.2 2 269
GenBank mRNA AY360471.1 - 136
AY596196.1 - 2 269
BC141249.1 - 2 267 9055861

View Full Product Details