Gene Transcripts
- Gene Symbol
- XPO7
- Entrez Gene ID
- 23039
- Gene Name
- exportin 7
- Gene Aliases
- EXP7, RANBP16
- Chromosome Location
- Chr. 8: 21919669 - 22006585 on Build GRCh38
- UniGene ID
- Hs.172685
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015024.4 |
NP_055839.3
|
2, 3 |
270 |
|
XM_017013244.1 |
XP_016868733.1
|
2, 3 |
497 |
|
XM_017013245.1 |
XP_016868734.1
|
2, 3 |
264 |
|
XM_017013246.1 |
XP_016868735.1
|
2, 3 |
493 |
|
XM_017013247.1 |
XP_016868736.1
|
2, 3 |
657 |
|
GenBank mRNA |
AB018288.1 |
- |
2, 3 |
180 |
|
AF064729.1 |
- |
2, 3 |
180 |
|
AK291472.1 |
- |
2, 3 |
185 |
|
AK311616.1 |
- |
2, 3 |
363 |
|
AL597247.1 |
- |
2, 3 |
226 |
|
BC030785.1 |
- |
2, 3 |
269 |
5269230 |
DB460798.1 |
- |
2, 3 |
272 |
|
DC332379.1 |
- |
2, 3 |
192 |
|
View Full Product Details