Gene Transcripts

Gene Symbol
ARL6IP1
Entrez Gene ID
23204
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 1
Gene Aliases
AIP1, ARL6IP, ARMER, SPG61
Chromosome Location
Chr. 16: 18791667 - 18801678 on Build GRCh38
UniGene ID
Hs.634882
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001313858.1 NP_001300787.1 6 1459
NM_015161.2 NP_055976.1 6 1535
GenBank mRNA AK300825.1 - 7 1343
BC010281.1 - 6 1392 3048642
BX648446.1 - 7 1322
D31885.1 - 6 1392

View Full Product Details