Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015631.5 | NP_056446.4 | 4 | 783 | |
XM_005269690.1 | XP_005269747.1 | 4 | 625 | ||
XM_011539627.1 | XP_011537929.1 | 4 | 625 | ||
XM_011539628.1 | XP_011537930.1 | 4 | 625 | ||
GenBank mRNA | AK075357.1 | - | 4 | 568 | |
AK098295.1 | - | 3 | 761 | ||
AK128834.1 | - | 4 | 1592 | ||
AL050022.1 | - | 4 | 567 | ||
AY359022.1 | - | 4 | 547 | ||
BC061582.1 | - | 4 | 454 | ||
BC068449.1 | - | 4 | 783 | 5298627 | |
DQ278872.1 | - | 4 | 783 |