Gene Transcripts

Gene Symbol
ASB3
Entrez Gene ID
51130
Gene Name
ankyrin repeat and SOCS box containing 3
Gene Aliases
ASB-3
Chromosome Location
Chr. 2: 53669980 - 53787009 on Build GRCh38
UniGene ID
Hs.40763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001201965.1 NP_001188894.1 2 644
NM_016115.4 NP_057199.1 3 478
NM_145863.2 NP_665862.1 2 269
GenBank mRNA AB209795.1 - 3 370
AF156778.1 - 2 276
AK000985.1 - 3 404
AK001283.1 - 2 202
AK056069.1 - 2 646
AK222595.1 - 2 288
AK222715.1 - 2 260
AK303133.1 - 3 404
BC006488.2 - 2 290 3537359
BC009569.1 - 3 336 4052577
DA038561.1 - 2 269

Gene Symbol
GPR75-ASB3
Entrez Gene ID
100302652
Gene Name
GPR75-ASB3 readthrough
Gene Aliases
Chromosome Location
Chr. 2: 53670293 - 53860033 on Build GRCh38
UniGene ID
Hs.40763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164165.1 NP_001157637.1 3 495
GenBank mRNA BC110915.1 - 3 417

View Full Product Details