Gene Transcripts

Gene Symbol
SHANK1
Entrez Gene ID
50944
Gene Name
SH3 and multiple ankyrin repeat domains 1
Gene Aliases
SPANK-1, SSTRIP, synamon
Chromosome Location
Chr. 19: 50659257 - 50719779 on Build GRCh38
UniGene ID
Hs.274255
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016148.3 NP_057232.2 5 700
XM_006723233.3 XP_006723296.1 6 1098
XM_011527013.2 XP_011525315.1 6 1098
XM_011527014.2 XP_011525316.1 6 1098
GenBank mRNA AF163302.1 - 5 700

View Full Product Details