Gene Transcripts

Gene Symbol
VPS36
Entrez Gene ID
51028
Gene Name
vacuolar protein sorting 36 homolog
Gene Aliases
C13orf9, CGI-145, EAP45
Chromosome Location
Chr. 13: 52412602 - 52450678 on Build GRCh38
UniGene ID
Hs.109520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282168.1 NP_001269097.1 8 632
NM_001282169.1 NP_001269098.1 8 793
NM_016075.3 NP_057159.2 8 659
GenBank mRNA AB208956.1 - 2 3333
AF151903.1 - 8 580
AK023182.1 - 6 376
AK289740.1 - 8 602
BC037279.1 - 8 608 4814999
BC050439.1 - 8 582 5764280
BX647847.1 - 8 600
CR933653.1 - 8 747

View Full Product Details