Gene Transcripts

Gene Symbol
CRNN
Entrez Gene ID
49860
Gene Name
cornulin
Gene Aliases
C1orf10, DRC1, PDRC1, SEP53
Chromosome Location
Chr. 1: 152409243 - 152414274 on Build GRCh38
UniGene ID
Hs.242057
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016190.2 NP_057274.1 3 1848
GenBank mRNA AF077831.1 - 3 1826
AK291301.1 - 3 1837
BC030807.1 - 3 1837 4271624

View Full Product Details