Gene Transcripts
- Gene Symbol
- ZNF44
- Entrez Gene ID
- 51710
- Gene Name
- zinc finger protein 44
- Gene Aliases
- GIOT-2, KOX7, ZNF, ZNF504, ZNF55, ZNF58
- Chromosome Location
- Chr. 19: 12224686 - 12294904 on Build GRCh38
- UniGene ID
- Hs.296731
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164276.1 |
NP_001157748.1
|
4, 5 |
536 |
|
NM_016264.3 |
NP_057348.3
|
3, 4 |
392 |
|
XM_011528062.2 |
XP_011526364.1
|
4, 5 |
501 |
|
XM_011528064.1 |
XP_011526366.1
|
3, 4 |
817 |
|
XM_011528066.1 |
XP_011526368.1
|
2, 3 |
690 |
|
XM_017026870.1 |
XP_016882359.1
|
3, 4 |
496 |
|
GenBank mRNA |
AB021642.1 |
- |
3, 4 |
327 |
|
AK297528.1 |
- |
4, 5 |
450 |
|
BC136502.1 |
- |
4, 5 |
431 |
9020491 |
CN364680.1 |
- |
3, 4 |
371 |
|
DA815414.1 |
- |
3, 4 |
295 |
|
View Full Product Details