Gene Transcripts
- Gene Symbol
- NEK2
- Entrez Gene ID
- 4751
- Gene Name
- NIMA related kinase 2
- Gene Aliases
- HsPK21, NEK2A, NLK1, PPP1R111, RP67
- Chromosome Location
- Chr. 1: 211658256 - 211675630 on Build GRCh38
- UniGene ID
- Hs.153704
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001204182.1 |
NP_001191111.1
|
2, 3 |
459 |
|
NM_001204183.1 |
NP_001191112.1
|
2, 3 |
459 |
|
NM_002497.3 |
NP_002488.1
|
2, 3 |
459 |
|
XM_005273147.1 |
XP_005273204.1
|
2, 3 |
456 |
|
GenBank mRNA |
AK223353.1 |
- |
2, 3 |
420 |
|
AK225522.1 |
- |
2, 3 |
420 |
|
AK303092.1 |
- |
3, 4 |
322 |
|
AY045701.1 |
- |
2, 3 |
390 |
|
BC043502.1 |
- |
2, 3 |
447 |
5541026 |
BC052807.1 |
- |
2, 3 |
454 |
|
BC065932.1 |
- |
2, 3 |
449 |
|
BT019729.1 |
- |
2, 3 |
308 |
|
DC407297.1 |
- |
2, 3 |
459 |
|
U11050.1 |
- |
2, 3 |
442 |
|
Z29066.1 |
- |
2, 3 |
390 |
|
View Full Product Details