Gene Transcripts

Gene Symbol
LETM1
Entrez Gene ID
3954
Gene Name
leucine zipper and EF-hand containing transmembrane protein 1
Gene Aliases
Chromosome Location
Chr. 4: 1811479 - 1856247 on Build GRCh38
UniGene ID
Hs.120165
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012318.2 NP_036450.1 3 656
XM_006713884.1 XP_006713947.1 3 364
XM_017008187.1 XP_016863676.1 2 777
GenBank mRNA AF061025.1 - 3 656
AK293572.1 - 3 364
AL133650.1 - 2 422
BC014500.1 - 3 556 4860194
BC021208.1 - 3 581 4126510

View Full Product Details