Gene Transcripts
- Gene Symbol
- TOR3A
- Entrez Gene ID
- 64222
- Gene Name
- torsin family 3 member A
- Gene Aliases
- ADIR, ADIR2
- Chromosome Location
- Chr. 1: 179081977 - 179095996 on Build GRCh38
- UniGene ID
- Hs.584957
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022371.3 |
NP_071766.2
|
2, 3 |
525 |
|
GenBank mRNA |
AJ299403.1 |
- |
2, 3 |
414 |
|
AJ299441.1 |
- |
2, 3 |
414 |
|
AK025998.1 |
- |
2, 3 |
404 |
|
BC001085.1 |
- |
2, 3 |
423 |
2959819 |
BC007571.1 |
- |
2, 3 |
414 |
3029304 |
BC011746.1 |
- |
2, 3 |
410 |
3343691 |
BC018292.1 |
- |
2, 3 |
415 |
3457259 |
CR457354.1 |
- |
2, 3 |
373 |
|
DA942161.1 |
- |
2, 3 |
525 |
|
View Full Product Details