Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152517.2 | NP_689730.2 | 1 | 1895 | |
GenBank mRNA | AK055552.1 | - | 2 | 1794 | |
AK296828.1 | - | 2 | 1730 | ||
BC033795.1 | - | 1 | 1879 | 5175299 | |
BX647620.1 | - | 1 | 1428 |