Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001272007.1 | NP_001258936.1 | 3 | 322 | |
NM_194279.3 | NP_919255.2 | 4 | 438 | ||
GenBank mRNA | AK290728.1 | - | 4 | 411 | |
BC015771.1 | - | 4 | 398 | 4849790 | |
BC032893.2 | - | 4 | 393 | 5259721 | |
BX114918.1 | - | 4 | 495 | ||
BX248252.1 | - | 4 | 396 | ||
DB501282.1 | - | 3 | 277 |