Gene Transcripts

Gene Symbol
RSL24D1
Entrez Gene ID
51187
Gene Name
ribosomal L24 domain containing 1
Gene Aliases
C15orf15, HRP-L30-iso, L30, RLP24, RPL24, RPL24L, TVAS3
Chromosome Location
Chr. 15: 55181314 - 55197033 on Build GRCh38
UniGene ID
Hs.274772
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016304.2 NP_057388.1 6 616
GenBank mRNA AB462502.1 - 6 500
AF060926.1 - 6 483
AF165521.1 - 6 616
AF201949.1 - 6 616
AF212226.1 - 6 616
AK055713.1 - 6 503
AK315431.1 - 6 516
BC008409.1 - 6 538 4097130
BC008422.1 - 6 508 4096889
BC008449.1 - 6 514 4133589
BC008499.1 - 6 529 4286115
BC009593.1 - 6 519 4109097
BC009604.1 - 6 519 4282059
BC012913.1 - 6 483 4514979
BC014576.1 - 6 518 3930303
BC016312.1 - 6 524 4096867
BC016331.1 - 6 524 4108672
BC016725.1 - 6 525 4078305
BC016777.1 - 6 506 4104810
BC026266.1 - 6 520 4276092
BC026267.1 - 6 499 4277062
BC028672.1 - 6 505 4092804
BC035995.1 - 6 529 4671856
BC093067.1 - 6 503 30419021
CR457179.1 - 6 473

View Full Product Details