Gene Transcripts

Gene Symbol
DCDC2
Entrez Gene ID
51473
Gene Name
doublecortin domain containing 2
Gene Aliases
DCDC2A, DFNB66, NPHP19, RU2, RU2S
Chromosome Location
Chr. 6: 24171755 - 24383292 on Build GRCh38
UniGene ID
Hs.61345
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195610.1 NP_001182539.1 2 490
NM_016356.4 NP_057440.2 1 583
GenBank mRNA AB032980.2 - 1 281
AF181721.1 - 1 583
AK027036.1 - 1 399
BC050704.1 - 2 490 5758845
BM923267.1 - 2 503

View Full Product Details