Gene Transcripts

Gene Symbol
PHF20
Entrez Gene ID
51230
Gene Name
PHD finger protein 20
Gene Aliases
C20orf104, GLEA2, HCA58, NZF, TDRD20A, TZP
Chromosome Location
Chr. 20: 35771999 - 35950377 on Build GRCh38
UniGene ID
Hs.517044
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016436.4 NP_057520.2 3 240
XM_011528843.1 XP_011527145.1 3 229
XM_017027864.1 XP_016883353.1 4 322
XM_017027865.1 XP_016883354.1 4 523
XM_017027866.1 XP_016883355.1 3 443
XM_017027867.1 XP_016883356.1 3 278
XM_017027868.1 XP_016883357.1 3 229
XM_017027869.1 XP_016883358.1 4 317
XM_017027870.1 XP_016883359.1 4 323
XM_017027871.1 XP_016883360.1 4 323
GenBank mRNA AF220416.1 - 3 204
AF258787.1 - 3 226
AF348207.1 - 2 165
AK054597.1 - 4 497
AK314503.1 - 3 204
AL558423.3 - 2 163
AY027523.1 - 2 165
AY134747.1 - 3 217
BC006415.1 - 3 180
BC016948.1 - 3 193
BC048210.1 - 4 273
BC080598.1 - 3 204
BC093405.1 - 3 240
BC150178.1 - 3 206 40148074
BG778940.1 - 4 310
BM906582.1 - 3 229
BX643875.1 - 3 198
CD690482.1 - 3 242
L09749.1 - 2 119

View Full Product Details