Gene Transcripts

Gene Symbol
UBAP1
Entrez Gene ID
51271
Gene Name
ubiquitin associated protein 1
Gene Aliases
NAG20, UAP, UBAP, UBAP-1
Chromosome Location
Chr. 9: 34179005 - 34252523 on Build GRCh38
UniGene ID
Hs.268963
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171201.1 NP_001164672.1 2 351
NM_001171203.2 NP_001164674.1 4 547
NM_001171204.2 NP_001164675.1 4 515
NM_016525.4 NP_057609.2 3 392
NR_033243.2 4 508
XM_006716779.3 XP_006716842.1 3 418
XM_011517899.1 XP_011516201.1 4 449
XM_017014801.1 XP_016870290.1 4 784
GenBank mRNA AF222043.2 - 3 321
AK074426.1 - 3 385
AK074724.1 - 1 130
AK074745.1 - 3 352
AK074812.1 - 3 392
AK074969.1 - 4 494
AK074995.1 - 3 356
AK223235.1 - 3 347
AK303711.1 - 2 351
AL136733.1 - 3 185
BC020950.1 - 3 354 3851813
BC098141.1 - 3 288 40011247
BC098316.1 - 3 288 40011248
BC099726.1 - 3 288 40011249
BC100668.1 - 3 288 40011243
BP237024.1 - 4 476
BX473128.1 - 4 500
CR533551.1 - 2 149
DA741347.1 - 4 502

View Full Product Details