Gene Transcripts
- Gene Symbol
- Clec16a
- Entrez Gene ID
- 74374
- Gene Name
- C-type lectin domain family 16, member A
- Gene Aliases
- 4932416N17Rik, curt, mKIAA0350
- Chromosome Location
- Chr. 16: 10545339 - 10744878 on Build GRCm38
- UniGene ID
- Mm.159176
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001204229.1 |
NP_001191158.1
|
16, 17 |
2034 |
|
NM_177562.5 |
NP_808230.2
|
17, 18 |
2040 |
|
GenBank mRNA |
AK016529.1 |
- |
7, 8 |
729 |
|
AK089327.1 |
- |
16, 17 |
2013 |
|
AK122255.1 |
- |
17, 18 |
1992 |
|
AK155314.1 |
- |
16, 17 |
2024 |
|
AK156927.1 |
- |
16, 17 |
1965 |
|
AK170754.1 |
- |
16, 17 |
2013 |
|
AK171134.1 |
- |
16, 17 |
2013 |
|
BC138907.1 |
- |
17, 18 |
1943 |
8861929 |
BC138909.1 |
- |
17, 18 |
1943 |
8861931 |
View Full Product Details