Gene Transcripts
- Gene Symbol
- Wdr46
- Entrez Gene ID
- 309628
- Gene Name
- WD repeat domain 46
- Gene Aliases
- Bing4
- Chromosome Location
- Chr. 20: 5447860 - 5455632 on Build Rnor_6.0
- UniGene ID
- Rn.104783
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_212491.2 |
NP_997656.1
|
11 |
1318 |
|
GenBank mRNA |
BC085934.1 |
- |
11 |
1318 |
|
View Full Product Details