Gene Transcripts

Gene Symbol
Stom
Entrez Gene ID
296655
Gene Name
stomatin
Gene Aliases
Chromosome Location
Chr. 3: 14515576 - 14538199 on Build Rnor_6.0
UniGene ID
Rn.229238
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011965.1 NP_001011965.1 6 607
GenBank mRNA BC083895.1 - 6 607

View Full Product Details