Gene Transcripts

Gene Symbol
Rpp40
Entrez Gene ID
291071
Gene Name
ribonuclease P/MRP 40 subunit
Gene Aliases
Chromosome Location
Chr. 17: 29638663 - 29647709 on Build Rnor_6.0
UniGene ID
Rn.23660
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013055.1 NP_001013073.1 8 1115
XM_006253790.1 XP_006253852.1 8 1077
GenBank mRNA BC091191.1 - 8 1115

View Full Product Details