Gene Transcripts

Gene Symbol
Akap8l
Entrez Gene ID
299569
Gene Name
A kinase (PRKA) anchor protein 8-like
Gene Aliases
Chromosome Location
Chr. 7: 14340168 - 14369831 on Build Rnor_6.0
UniGene ID
Rn.107308
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013946.1 NP_001013968.1 11 1427
XM_006241054.1 XP_006241116.1 12 1335
XM_006241055.1 XP_006241117.1 11 1437
GenBank mRNA BC085785.1 - 11 1427

View Full Product Details