Gene Transcripts

Gene Symbol
Bles03
Entrez Gene ID
266609
Gene Name
basophilic leukemia expressed protein BLES03
Gene Aliases
Chromosome Location
Chr. 1: 220806563 - 220808817 on Build Rnor_6.0
UniGene ID
Rn.7405
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024233.2 NP_001019404.2 2 853
NM_001243534.1 NP_001230463.1 2 867
GenBank mRNA AF502571.2 - 1 60
BC093387.1 - 2 712 7389419
CK483724.1 - 2 709
FQ233132.1 - 2 746

View Full Product Details