Gene Transcripts

Gene Symbol
Ube2l6
Entrez Gene ID
295704
Gene Name
ubiquitin-conjugating enzyme E2L 6
Gene Aliases
Chromosome Location
Chr. 3: 72191533 - 72206190 on Build Rnor_6.0
UniGene ID
Rn.8609
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024755.1 NP_001019926.1 4 1211
GenBank mRNA BC097364.1 - 4 1211
FQ220881.1 - 4 813
FQ227712.1 - 4 783

View Full Product Details