Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017721.4 | NP_060191.3 | 6 | 853 | |
XM_005259972.2 | XP_005260029.1 | 6 | 801 | ||
XM_005259973.2 | XP_005260030.1 | 6 | 801 | ||
XM_005259974.2 | XP_005260031.1 | 6 | 801 | ||
XM_005259975.2 | XP_005260032.1 | 6 | 801 | ||
GenBank mRNA | AB097002.1 | - | 6 | 795 | |
AF536205.1 | - | 8 | 906 | ||
AK000248.1 | - | 373 | |||
AK123155.1 | - | 4 | 611 | ||
AK315068.1 | - | 6 | 771 | ||
BC006556.2 | - | 2 | 91 | ||
BC064981.1 | - | 6 | 757 | 6585236 | |
BC113968.1 | - | 5 | 482 |