Gene Transcripts

Gene Symbol
RGAG4
Entrez Gene ID
340526
Gene Name
retrotransposon gag domain containing 4
Gene Aliases
6430402L03Rik, MAR5, MART5
Chromosome Location
Chr. X: 72127110 - 72131901 on Build GRCh38
UniGene ID
Hs.512180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024455.3 NP_001019626.1 1 590
GenBank mRNA AB082532.1 - 1 590
BC151224.1 - 1 611 9007468

View Full Product Details