Gene Transcripts

Gene Symbol
Krtap31-2
Entrez Gene ID
432602
Gene Name
keratin associated protein 31-2
Gene Aliases
OTTMUSG00000002177
Chromosome Location
Chr. 11: 99936291 - 99937225 on Build GRCm38
UniGene ID
Mm.246878
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025244.3 NP_001020415.1 1 918
GenBank mRNA AK037252.1 - 1 921

View Full Product Details