Gene Transcripts

Gene Symbol
Reep4
Entrez Gene ID
306014
Gene Name
receptor accessory protein 4
Gene Aliases
RGD1306561
Chromosome Location
Chr. 15: 52235283 - 52238490 on Build Rnor_6.0
UniGene ID
Rn.6988
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025279.1 NP_001020450.1 3 142
GenBank mRNA BC097957.1 - 3 142
FQ234004.1 - 4 369

View Full Product Details