Gene Transcripts

Gene Symbol
STAG3L2
Entrez Gene ID
442582
Gene Name
stromal antigen 3-like 2 (pseudogene)
Gene Aliases
STAG3L1, STAG3L2P, STAG3L3
Chromosome Location
Chr. 7: 74881990 - 74890612 on Build GRCh38
UniGene ID
Hs.632310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040584.2 8 1465
GenBank mRNA BM311130.1 - 1 87

Gene Symbol
STAG3L1
Entrez Gene ID
54441
Gene Name
stromal antigen 3-like 1 (pseudogene)
Gene Aliases
STAG3L1P, STAG3L2, STAG3L3
Chromosome Location
Chr. 7: 75359200 - 75367809 on Build GRCh38
UniGene ID
Hs.632310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040583.2 10 1721

Gene Symbol
STAG3L3
Entrez Gene ID
442578
Gene Name
stromal antigen 3-like 3 (pseudogene)
Gene Aliases
STAG3L1, STAG3L2, STAG3L3P
Chromosome Location
Chr. 7: 72997295 - 73005943 on Build GRCh38
UniGene ID
Hs.632310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040582.2 9 1740
GenBank mRNA BC030509.1 - 5 1017

View Full Product Details