Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003685.3 | NP_001003685.2 | 13 | 1274 | |
GenBank mRNA | AK030504.1 | - | 13 | 1227 | |
BC120748.1 | - | 13 | 1204 | 40129671 | |
BC120774.1 | - | 13 | 1204 | 40129697 | |
BC144892.1 | - | 13 | 1215 | ||
L07379.1 | - | 13 | 1207 |