Gene Transcripts

Gene Symbol
KRTAP5-2
Entrez Gene ID
440021
Gene Name
keratin associated protein 5-2
Gene Aliases
KRTAP5-8, KRTAP5.2
Chromosome Location
Chr. 11: 1597177 - 1598294 on Build GRCh38
UniGene ID
Hs.633229
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004325.1 NP_001004325.1 1 216
GenBank mRNA AB126071.1 - 1 224
AJ628245.1 - 1 216
BC132882.1 - 1 195 40146904
BC132908.1 - 1 195 40146930

View Full Product Details