Gene Transcripts

Gene Symbol
OR2M2
Entrez Gene ID
391194
Gene Name
olfactory receptor family 2 subfamily M member 2
Gene Aliases
OR2M2Q, OST423
Chromosome Location
Chr. 1: 248179986 - 248181029 on Build GRCh38
UniGene ID
Hs.534728
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004688.1 NP_001004688.1 1 941

View Full Product Details