Gene Transcripts

Gene Symbol
Vom1r4
Entrez Gene ID
494236
Gene Name
vomeronasal 1 receptor 4
Gene Aliases
V1re24
Chromosome Location
Chr. 1: 86533127 - 86534023 on Build Rnor_6.0
UniGene ID
Rn.129461
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009516.1 NP_001009516.1 1 505
GenBank mRNA AY510338.1 - 1 505

View Full Product Details