Gene Transcripts

Gene Symbol
Aprt
Entrez Gene ID
292072
Gene Name
adenine phosphoribosyl transferase
Gene Aliases
Chromosome Location
Chr. 19: 55387288 - 55389256 on Build Rnor_6.0
UniGene ID
Rn.2498
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013061.1 NP_001013079.1 4 335
GenBank mRNA FQ218973.1 - 362
FQ228571.1 - 4 353

View Full Product Details