Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013657.2 | NP_001013679.2 | 4 | 200 | |
NM_001322811.1 | NP_001309740.1 | 5 | 319 | ||
GenBank mRNA | AK130527.1 | - | 4 | 786 | |
BC127821.1 | - | 4 | 229 | ||
BX440248.2 | - | 4 | 200 |