Gene Transcripts

Gene Symbol
Lpin3
Entrez Gene ID
362261
Gene Name
lipin 3
Gene Aliases
Chromosome Location
Chr. 3: 156874812 - 156899438 on Build Rnor_6.0
UniGene ID
Rn.145057
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014184.1 NP_001014206.1 10, 11 1521
XM_006235487.2 XP_006235549.1 12, 13 2946
XM_006235489.2 XP_006235551.1 12, 13 2297
XM_006235490.2 XP_006235552.1 13, 14 3091
XM_006235491.2 XP_006235553.1 12, 13 2557
XM_006235493.2 XP_006235555.1 11, 12 2091
XM_006235494.2 XP_006235556.1 11, 12 2740
GenBank mRNA BC089878.1 - 10, 11 1521

View Full Product Details