Gene Transcripts

Gene Symbol
Wrap73
Entrez Gene ID
366515
Gene Name
WD repeat containing, antisense to TP73
Gene Aliases
Wdr8
Chromosome Location
Chr. 5: 171441536 - 171455795 on Build Rnor_6.0
UniGene ID
Rn.30003
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014262.1 NP_001014284.1 6, 7 638
GenBank mRNA BC082062.1 - 6, 7 638

View Full Product Details