Gene Transcripts

Gene Symbol
C20orf27
Entrez Gene ID
54976
Gene Name
chromosome 20 open reading frame 27
Gene Aliases
Chromosome Location
Chr. 20: 3753499 - 3768388 on Build GRCh38
UniGene ID
Hs.274422
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039140.2 NP_001034229.1 3, 4 474
NM_001258429.1 NP_001245358.1 3, 4 399
NM_001258430.1 NP_001245359.1 3, 4 853
NR_047675.1 3, 4 543
XM_011529266.2 XP_011527568.1 3, 4 454
GenBank mRNA AK000557.1 - 3, 4 338
AK024220.1 - 3, 4 338
AK097645.1 - 3, 4 981
AK124341.1 - 3, 4 404
AK125202.1 - 1, 2 1307
AK225109.1 - 3, 4 338
AK290955.1 - 3, 4 543
AK308520.1 - 3, 4 338
BC012196.1 - 3, 4 325 2822218
BC024036.1 - 3, 4 336 3510659
BF978883.1 - 3, 4 400
BG474804.1 - 3, 4 393

View Full Product Details