Gene Transcripts

Gene Symbol
BCAS4
Entrez Gene ID
55653
Gene Name
breast carcinoma amplified sequence 4
Gene Aliases
CNOL
Chromosome Location
Chr. 20: 50794894 - 50877736 on Build GRCh38
UniGene ID
Hs.381178
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010974.2 NP_001010974.1 3 411
NM_017843.4 NP_060313.3 3 411
NM_198799.3 NP_942094.2 3 411
XM_011528886.2 XP_011527188.1 3 334
XM_011528887.2 XP_011527189.1 3 334
XM_017027932.1 XP_016883421.1 3 334
GenBank mRNA AF361220.1 - 3 411
AJ511266.1 - 3 235
AK000502.1 - 3 274
BC038381.1 - 3 261 4472674
BC047337.1 - 3 240
BP212127.1 - 3 375

View Full Product Details