Gene Transcripts

Gene Symbol
Ier5l
Entrez Gene ID
499772
Gene Name
immediate early response 5-like
Gene Aliases
Chromosome Location
Chr. 3: 9036351 - 9037942 on Build Rnor_6.0
UniGene ID
Rn.162656
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025041.1 NP_001020212.1 1 1266
GenBank mRNA BC087095.1 - 1 1266

View Full Product Details