Gene Transcripts

Gene Symbol
TSPAN17
Entrez Gene ID
26262
Gene Name
tetraspanin 17
Gene Aliases
FBX23, FBXO23, TM4SF17
Chromosome Location
Chr. 5: 176647387 - 176659058 on Build GRCh38
UniGene ID
Hs.532129
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006616.2 NP_001006617.2 9 1743
NM_012171.2 NP_036303.1 9 1784
NM_130465.4 NP_569732.2 9 1775
XM_005265876.3 XP_005265933.1 7 1557
GenBank mRNA AK024427.1 - 8 3646
AY037146.1 - 4 2010
BC010405.2 - 9 1565 3621871
BC067105.1 - 9 1660 6646576
BG703362.1 - 1 265
CT002861.1 - 1 645

View Full Product Details